missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Alexa Fluor™ 647 Alkyne, Triethylammonium Salt DFS Item

Alexa Fluor™ 647 alkyne, triethylammonium salt

Supplier:  Molecular Probes™ A10278

Catalog No. A10278


Only null left
Explore available promotions

Description

Description

The far red-fluorescent Alexa Fluor™ 647 alkyne is reactive with azides via a copper-catalyzed click reaction. The bright and photostable fluorophore can be for used with flow cytometry, microscopy and HCS
Specifications

Specifications

Solid
Alkyne
Store at ≤-20°C, desiccated and protect from light.
Fluorescence
Room Temperature
Alexa Fluor™ 647
Far-red
0.5 mg
DMSO (Dimethylsulfoxide)
Azide
Alkyne
Alkyne
Alexa Fluor
Alexa Fluor, Molecular Probes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.