missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ ALCALASE™ Enzyme, Bacillus licheniformis

Catalog No. 126741500ML
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
126741500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 126741500ML Supplier MilliporeSigma™ Supplier No. 126741500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Suitable for hydrolysis of proteins

  • Serine endopeptidase that consists primarily of subtilisin A
  • Density: 1.25g/mL

Specifications

pH Range Optimum 7.0 to 8.5
Content And Storage 2°C to 8°C
Form Liquid
Quantity 500 mL
Product Type Alcalase Enzyme, Bacillus licheniformis