missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alanine, USP, 98.5-101.5%, Spectrum™ Chemical
A1253, 56-41-7, C3H7NO2
$371.90 - $11,253.19
Chemical Identifiers
| CAS | 56-41-7 |
|---|---|
| Molecular Formula | C3H7NO2 |
| Molecular Weight (g/mol) | 89.09 |
| MDL Number | MFCD00064410 |
| InChI Key | QNAYBMKLOCPYGJ-REOHCLBHSA-N |
| IUPAC Name | (2S)-2-aminopropanoic acid |
| SMILES | C[C@H](N)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18600341
|
N/A | 100 g | Amber Glass Bottle |
Each for $371.90
|
|
||||
|
18600342
|
N/A | 1 kg | Amber Glass Bottle |
Each for $1,362.29
|
|
||||
|
18600343
|
N/A | 5 kg | Poly Pail |
Each for $4,805.13
|
|
||||
|
18600344
|
N/A | 25 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Alanine, USP is a non-essential alpha-amino acid and appears as a white powder. Alanine is used in clinical nutrition and as an ingredient for pharmaceutical products such as therapeutics for prostate hypertrophy. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 56-41-7 | |
| 89.09 | |
| QNAYBMKLOCPYGJ-REOHCLBHSA-N | |
| C[C@H](N)C(O)=O |
| C3H7NO2 | |
| MFCD00064410 | |
| (2S)-2-aminopropanoic acid |
Specifications
| 56-41-7 | |
| 0.05% | |
| 30 ppm | |
| C3H7NO2 | |
| +13.7° to +15.1° | |
| QNAYBMKLOCPYGJ-REOHCLBHSA-N | |
| (2S)-2-aminopropanoic acid | |
| 98.5 to 101.5% |
| 100% | |
| 0.15% | |
| 0.2% | |
| MFCD00064410 | |
| 0.03% | |
| C[C@H](N)C(O)=O | |
| 89.09 | |
| USP |