missing translation for 'onlineSavingsMsg'
Learn More

Alanine, USP, 98.5-101.5%, Spectrum™ Chemical

Catalog No. 18600341
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600341 100 g Amber Glass Bottle
18600342 1 kg Amber Glass Bottle
18600343 5 kg Poly Pail
18600344 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18600341 Supplier Spectrum Chemical Mfg Cor Supplier No. A1253100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

A1253, 56-41-7, C3H7NO2

Spectrum™ Chemical Alanine, USP is a non-essential alpha-amino acid and appears as a white powder. Alanine is used in clinical nutrition and as an ingredient for pharmaceutical products such as therapeutics for prostate hypertrophy. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 56-41-7
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00064410
InChI Key QNAYBMKLOCPYGJ-REOHCLBHSA-N
IUPAC Name (2S)-2-aminopropanoic acid
SMILES C[C@H](N)C(O)=O

Specifications

CAS Max % 100%
Chloride 0.05%
Ignition Residue 0.15%
Iron (Fe) 30 ppm
Loss on Drying 0.2%
Packaging Amber Glass Bottle
Specific Rotation +13.7° to +15.1°
Sulfate 0.03%
Quantity 100 g
Percent Purity 98.5 to 101.5%
Grade USP
Show More Show Less