Learn More
Applied Biosystems™ AgPath-ID™ One-Step RT-PCR Reagents with manual
AgPath-ID™ One-Step RT-PCR Reagents are designed for sensitive, robust amplification of RNA targets using a rapid single-tube TaqMan™ real-time reverse transcription PCR (RT-PCR) strategy.
$399.00 - $3,155.00
Specifications
| Concentration | 25X |
|---|---|
| Description | Works on all AB platforms, but is optimized for the 7500. |
| Detection Method | Primer-probe |
| Format | Tube |
| GC-Rich PCR Performance | High |
| Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
4387424M
|
Applied Biosystems™
4387424M |
500 Reactions |
Each for $1,850.00
|
|
|||||
4387391M
|
Applied Biosystems™
4387391M |
1000 Reactions |
Each for $3,155.00
|
|
|||||
AM1005M
|
Applied Biosystems™
AM1005M |
100 Reactions |
Each for $399.00
|
|
|||||
Description
- Recommended for RNA pathogen amplification
- Consistent amplification of RNA targets with high specificity and sensitivity
- Optimized to work with your target-specific primers and probes
- Contains ROX for quantitative fluorescent signal normalization
AgPath-ID™ One-Step RT-PCR Reagents are configured for fast and simple reaction setup. The reactions are assembled in a single tube, minimizing sample handling errors and expediting set-up time. Once assembled, results are available in approximately one hour. The 25X RT-PCR Enzyme Mix included in the kit contains highly efficient ArrayScript™ Reverse Transcriptase, a mutant MMLV RT that produces high cDNA yields, and AmpliTaq Gold™ polymerase, the preferred hot-start DNA polymerase for specific target amplification. The 2X RT-PCR Buffer has been optimized for efficient, robust reverse transcription and PCR includes the passive reference dye, ROX™ Dye, for quantitative fluorescent signal normalization.
Specifications
| 25X | |
| Primer-probe | |
| High | |
| AmpliTaq Gold DNA Polymerase | |
| 7500 System | |
| AgPath-ID, Ambion | |
| ArrayScript™ |
| Works on all AB platforms, but is optimized for the 7500. | |
| Tube | |
| 1-step RT-qPCR | |
| Standard | |
| ROX (Pre-mixed) | |
| One-Step RT-PCR Reagent | |
| 25 X |
For Research Use Only. Not for use in diagnostic procedures.