missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ AGI-6780, 95%

Catalog No. AC467821000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467821000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC467821000 Supplier Thermo Scientific™ Supplier No. 467821000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material AGI-6780
CAS 1432660-47-3
Synonym N-Cyclopropyl-4-(3-thienyl)-3-[[[[3-(trifluoromethyl)phenyl]amino]carbonyl]amino]-benzenesulfonamide
Quantity 100 mg
Molecular Formula C21 H18 F3 N3 O3 S2
Formula Weight 481.51
Physical Form Solid
Product Type Reagent
Purity 95%