missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AGI 5198

Catalog No. 708725
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
708725 25 mg
70875 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 708725 Supplier Tocris Bioscience™ Supplier No. 7087/25*
Only null left

Potent and selective inhibitor of mutant isocitrate dehydrogenase 1 (mIDH1)

AGI 5198 is a potent and selective inhibitor of mutant isocitrate dehydrogenase 1 (mIDH1; IC50 = 70 nM). Selective for mIDH1 over wild-type IDH1 and IDH2. Inhibits production of oncometabolite D-2 hydroxyglutarate. Attenuates growth of glioma cells containing mIDH1, but not wild-type IDH1, in vitro and in vivo. Also inhibits growth of fibrosarcoma and chondrosarcoma cells bearing mIDH1.

Specifications

Chemical Name or Material N-[2-(Cyclohexylamino)-1-(2-methylphenyl)-2-oxoethyl]-N-(3-fluorophenyl)-2-methyl-1 H-Imidazole-1-acetamide
CAS 1355326-35-0
Quantity 25 mg
Target Dehydrogenase Inhibitors
Molecular Formula C27H31FN4O2
Purity 0.98