missing translation for 'onlineSavingsMsg'
Learn More

Agarose (Protein Electrophoresis Grade), Fisher BioReagents™

$186.61 - $1,546.82

Chemical Identifiers

CAS 9012-36-6
Molecular Formula C12H18O9
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
BP16425
View Documents Promotion Details
Fisher BioReagents
FLBP164 25
25 g Poly Bottle
Each for $186.61
Case of 6 Each for $1,119.66
Only null left
 
BP164100
View Documents Promotion Details
Fisher BioReagents
BP164 100
100 g Poly Bottle
Each for $523.18
Case of 6 Each for $3,139.08
Only null left
 
BP164500
View Documents Promotion Details
Fisher BioReagents
BP164500
500 g Poly Bottle
Each for $1,546.82
Only null left
 
Description

Description

Agarose (Protein Electrophoresis Grade)

Specifications

Chemical Identifiers

9012-36-6
C12H18O9

Specifications

Powder/Solid
Cream
0.1 max.
800g/cm2min.
306.12
Agarose
9012-36-6
DNase free
34° to 45°C
C12H18O9
DNase- and RNase-Free
Videos
Safety and Handling

Safety and Handling

Recommended Storage : RT

SDS
Documents

Documents