missing translation for 'onlineSavingsMsg'
Learn More

Agarose (Protein Electrophoresis Grade), Fisher BioReagents™

Catalog No. BP164500
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP164500 500 g Poly Bottle
BP16425 25 g Poly Bottle
BP164100 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP164500 Supplier Fisher BioReagents Supplier No. BP164500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 9012-36-6 | Molecular Formula: C12H18O9

Agarose (Protein Electrophoresis Grade)

Chemical Identifiers

CAS 9012-36-6
Molecular Formula C12H18O9

Specifications

Physical Form Powder/Solid
Color Cream
Quantity 500 g
DNase DNase free
Electroendosmosis (EEO) 0.1 max.
Gelation Temperature 34° to 45°C
Gel Strength 800g/cm2min.
Packaging Poly Bottle
Formula Weight 306.12
Grade DNase- and RNase-Free
Chemical Name or Material Agarose
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT