missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agarose (Protein Electrophoresis Grade), Fisher BioReagents™
Supplier: Fisher BioReagents BP164 100
| Quantity | 100 g |
|---|---|
| Packaging | Poly Bottle |
Description
Agarose (Protein Electrophoresis Grade)
Chemical Identifiers
| 9012-36-6 | |
| C12H18O9 |
Specifications
| Agarose | |
| 9012-36-6 | |
| 100 g | |
| 0.1 max. | |
| 800g/cm2min. | |
| DNase- and RNase-Free | |
| 306.12 |
| Powder/Solid | |
| Cream | |
| DNase free | |
| 34° to 45°C | |
| Poly Bottle | |
| C12H18O9 | |
| Protein Electrophoresis |
Safety and Handling
Recommended Storage : RT