missing translation for 'onlineSavingsMsg'
Learn More

Agarose, Electrophoresis Grade, MP Biomedicals™

Electroendosmosis: 0.13; Gel Strength: ≥600 gm/cm2; Gel Temperature: 36°C; This is a standard agarose with good gel strength and clarity, suitable for all general applications

$536.99 - $1807.86

Chemical Identifiers

CAS 9012-36-6
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN820721
View Documents
MP Biomedicals Inc
04820721
100 g
Each for $536.99
Only null left
 
ICN820723
View Documents
MP Biomedicals Inc
04820723
500 g
Each for $1,807.86
Only null left
 
Description

Description

Specifications

Chemical Identifiers

9012-36-6

Specifications

9012-36-6
100 g
Electrophoresis
Agarose
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'