Learn More
Agarose, Electrophoresis Grade, MP Biomedicals™
Electroendosmosis: 0.13; Gel Strength: ≥600 gm/cm2; Gel Temperature: 36°C; This is a standard agarose with good gel strength and clarity, suitable for all general applications
Supplier:  MP Biomedicals Inc 04820723
 View more versions of this product
Catalog No. ICN820723
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
Agarose
9012-36-6
500 g
Electrophoresis
Chemical Identifiers
9012-36-6
Product Suggestions
Product Suggestions