Learn More
Agarose, Electrophoresis Grade, MP Biomedicals™
Electroendosmosis: 0.13; Gel Strength: ≥600 gm/cm2; Gel Temperature: 36°C; This is a standard agarose with good gel strength and clarity, suitable for all general applications
Supplier: MP Biomedicals Inc 04820721
Specifications
| Agarose | |
| 9012-36-6 | |
| 100 g | |
| Electrophoresis |
Chemical Identifiers
| 9012-36-6 |