missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agar, Powder, FCC, Spectrum™ Chemical
A1672, 9002-18-0
$397.21 - $21198.68
Chemical Identifiers
| CAS | 9002-18-0 |
|---|---|
| Molecular Formula | C14H24O9 |
| Molecular Weight (g/mol) | 336.34 |
| InChI Key | GYYDPBCUIJTIBM-UHFFFAOYNA-N |
| IUPAC Name | 2-({4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl}oxy)-6-(hydroxymethyl)-4-methoxyoxane-3,5-diol |
| SMILES | COC1C(O)C(CO)OC(OC2C3COC2C(O)C(C)O3)C1O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18600331
|
Spectrum Chemical Mfg Cor
A1672125GM |
125 g | Poly Bottle |
Each for $397.21
|
|
||||
|
18600332
|
Spectrum Chemical Mfg Cor
A1672500GM |
500 g | Poly Bottle |
Each for $1,036.54
|
|
||||
|
18600333
|
Spectrum Chemical Mfg Cor
A167225KG |
2.5 kg | Poly Bottle |
Each for $4,197.32
|
|
||||
|
18600334
|
Spectrum Chemical Mfg Cor
A167225KGBL |
25 kg | Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Agar, Powder, FCC is a gel made from the cell walls of several species of red algae or seaweed. It appears as an odorless, tasteless, beige powder. It functions as a binder and thickener in cooking to make jellies, pudding, and custards. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Chemical Identifiers
| 9002-18-0 | |
| 336.34 | |
| 2-({4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl}oxy)-6-(hydroxymethyl)-4-methoxyoxane-3,5-diol |
| C14H24O9 | |
| GYYDPBCUIJTIBM-UHFFFAOYNA-N | |
| COC1C(O)C(CO)OC(OC2C3COC2C(O)C(C)O3)C1O |
Specifications
| 9002-18-0 | |
| Poly Bottle | |
| Beige to Brown | |
| GYYDPBCUIJTIBM-UHFFFAOYNA-N | |
| 2-({4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl}oxy)-6-(hydroxymethyl)-4-methoxyoxane-3,5-diol | |
| FCC |
| 100% | |
| C14H24O9 | |
| 125 g | |
| COC1C(O)C(CO)OC(OC2C3COC2C(O)C(C)O3)C1O | |
| 336.34 | |
| Agar |