missing translation for 'onlineSavingsMsg'
Learn More

Agar, Powder, FCC, Spectrum™ Chemical

A1672, 9002-18-0

Supplier:  Spectrum Chemical Mfg Cor A167225KG

 View more versions of this product

Catalog No. 18600333

Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Agar, Powder, FCC is a gel made from the cell walls of several species of red algae or seaweed. It appears as an odorless, tasteless, beige powder. It functions as a binder and thickener in cooking to make jellies, pudding, and custards. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

Agar
100%
C14H24O9
2.5 kg
COC1C(O)C(CO)OC(OC2C3COC2C(O)C(C)O3)C1O
336.34
9002-18-0
Poly Bottle
Beige to Brown
GYYDPBCUIJTIBM-UHFFFAOYNA-N
2-({4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl}oxy)-6-(hydroxymethyl)-4-methoxyoxane-3,5-diol
FCC

Chemical Identifiers

9002-18-0
336.34
2-({4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl}oxy)-6-(hydroxymethyl)-4-methoxyoxane-3,5-diol
C14H24O9
GYYDPBCUIJTIBM-UHFFFAOYNA-N
COC1C(O)C(CO)OC(OC2C3COC2C(O)C(C)O3)C1O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions