missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ AFM Primer
Specifications
Specifications
| Content And Storage | -20°C. Avoid freeze/thaw cycles. |
| Product Line | Novus |
| Quantity | 0.2 mL |
| For Use With (Application) | ChIP |
| Type | Primer |