missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Afatinib

Catalog No. AC465640010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465640010 1 g
AC465642500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465640010 Supplier Thermo Scientific Chemicals Supplier No. 465640010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 850140-72-6
Molecular Formula C24H25ClFN5O3

Specifications

Quantity 1 g
Melting Point 102.0°C
Formula Weight 485.94
InChI Key ULXXDDBFHOBEHA-UHFFFAOYNA-N
SMILES CN(C)CC=CC(=O)NC1=C(OC2CCOC2)C=C2N=CN=C(NC3=CC(Cl)=C(F)C=C3)C2=C1
IUPAC Name N-{4-[(3-chloro-4-fluorophenyl)amino]-7-(oxolan-3-yloxy)quinazolin-6-yl}-4-(dimethylamino)but-2-enamide
Molecular Weight (g/mol) 485.94
Chemical Name or Material Afatinib
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

If eye irritation persists: Get medical advice/attention.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Wear protective gloves/protective clothing/eye protection/face protection.

If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only