missing translation for 'onlineSavingsMsg'
Learn More

Adonitol, 99.8%, MP Biomedicals™

Catalog No. ICN10025225
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10025225 25 g
ICN10025280 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10025225 Supplier MP Biomedicals Inc Supplier No. 0210025225

Chemical Identifiers

CAS 488-81-3
Molecular Formula C5H12O5
Molecular Weight (g/mol) 152.146
MDL Number MFCD00064291
InChI Key HEBKCHPVOIAQTA-NGQZWQHPSA-N
Synonym xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
PubChem CID 6912
IUPAC Name (2S,4R)-pentane-1,2,3,4,5-pentol
SMILES C(C(C(C(CO)O)O)O)O

Specifications

Color White
Density 1.53g/cm3
Assay Percent Range 0.998
Quantity 25 g
Formula Weight 152.15
Physical Form Powder
Chemical Name or Material Adonitol
Recommended Storage Store at 4°C.