missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adonitol, 98%

Catalog No. AC167030050
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC167030050 5 g Glass bottle
AC167030100 10 g Glass bottle
AC167031000 100 g Glass bottle
AC167035000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC167030050 Supplier Thermo Scientific Chemicals Supplier No. 167030050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Adonitol, 98%, C5H12O5, CAS Number-488-81-3, adonit, adonite, adonitol, d-ribitol, d-xylitol, eutrit, ribitol, xylit, xylite, xylitol, 100g, 97.5% min. (ELSD) (HPLC), 01,53, 100.0, 99.0, White, 207-685-7, 152.15, HEBKCHPVOIAQTA-NGQZWQHPSA-N, Authentic, (2S,4R)-pentane-1,2,3,4,5-pentol | CAS: 488-81-3 | C5H12O5 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 488-81-3
Molecular Formula C5H12O5
Molecular Weight (g/mol) 152.15
MDL Number MFCD00064291
InChI Key HEBKCHPVOIAQTA-NGQZWQHPSA-N
Synonym xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
PubChem CID 6912
IUPAC Name (2S,4R)-pentane-1,2,3,4,5-pentol
SMILES C(C(C(C(CO)O)O)O)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 101.0°C to 104.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 01,53
Merck Index 15,157
Solubility Information Solubility in water: soluble. Other solubilities: soluble in hot alcohol,insoluble in ether
Formula Weight 152.15
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material Adonitol
Show More Show Less
EINECSNumber 207-685-7

RUO – Research Use Only