missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ AdipoRon hydrochloride

Catalog No. 509650
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
509650 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 509650 Supplier R&D Systems™ Supplier No. 5096/50

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Tocris product, part of R&D Systems. Orally active AdipoR1 and AdipoR2 agonist; antidiabetic

Agonist of adiponectin receptors AdipoR1 and AdipoR2 (Kd values are 1.8 and 3.1 μM respectively). Activates AMPK and PPARα signaling; ameliorates insulin resistance, dyslipidemia and glucose intolerance in db/db mice. Orally active antidiabetic agent.

Chemical Identifiers

CAS 1781835-20-8
Molecular Formula C27H29ClN2O3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 464.98
Molecular Weight (g/mol) 464.99
Chemical Name or Material AdipoRon hydrochloride
Recommended Storage Desiccate at RT