missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenosine 3',5'-cyclic monophosphate, 99+%

CAS: 60-92-4 | C10H12N5O6P | 329.21 g/mol

$1,939.73

Chemical Identifiers

CAS 60-92-4
Molecular Formula C10H12N5O6P
Molecular Weight (g/mol) 329.21
MDL Number MFCD00005845
InChI Key IVOMOUWHDPKRLL-YPLCUDRINA-N
Synonym camp, cyclic amp, adenosine 3',5'-cyclic monophosphate, 3',5'-cyclic amp, adenosine 3',5'-phosphate, adenosine 3',5'-cyclophosphate, cyclic 3',5'-amp, adenosine cyclic monophosphate, adenosine-3',5'-cyclophosphate, cyclic adenosine monophosphate
PubChem CID 6076
ChEBI CHEBI:17489
SMILES NC1=C2N=CN([C@@H]3O[C@@H]4COP(O)(=O)O[C@H]4[C@H]3O)C2=NC=N1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC225800050
View Documents Promotion Details
Thermo Scientific Chemicals
225800050
5 g Glass bottle
Each for $1,939.73
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

60-92-4
329.21
IVOMOUWHDPKRLL-YPLCUDRINA-N
6076
NC1=C2N=CN([C@@H]3O[C@@H]4COP(O)(=O)O[C@H]4[C@H]3O)C2=NC=N1
C10H12N5O6P
MFCD00005845
camp, cyclic amp, adenosine 3',5'-cyclic monophosphate, 3',5'-cyclic amp, adenosine 3',5'-phosphate, adenosine 3',5'-cyclophosphate, cyclic 3',5'-amp, adenosine cyclic monophosphate, adenosine-3',5'-cyclophosphate, cyclic adenosine monophosphate
CHEBI:17489

Specifications

60-92-4
lambda max. 260nm,pH 7,0.77 to 0.83 (A250/A260),0.13 to 0.19 (A280/A260)
260.0°C
C10H12N5O6P
26, IV, 3618
− 53.70
Solubility in water: 50mg/mL
NC1=C2N=CN([C@@H]3O[C@@H]4COP(O)(=O)O[C@H]4[C@H]3O)C2=NC=N1
329.21
CHEBI:17489
99+%
99.0
100.0
99% min. (HPLC)
MFCD00005845
15, 2701
camp, cyclic amp, adenosine 3',5'-cyclic monophosphate, 3',5'-cyclic amp, adenosine 3',5'-phosphate, adenosine 3',5'-cyclophosphate, cyclic 3',5'-amp, adenosine cyclic monophosphate, adenosine-3',5'-cyclophosphate, cyclic adenosine monophosphate
IVOMOUWHDPKRLL-YPLCUDRINA-N
(4aR,6R,7R,7aS)-6-(6-aminopurin-9-yl)-2-hydroxy-2-oxo-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-7-ol
6076
329.2
Adenosine 3′,5′-cyclic monophosphate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 200-492-9

SDS
Documents

Documents

RUO – Research Use Only