missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenosine-3',5'-cyclic monophosphate, 98%

Catalog No. AAJ6093603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6093603 1 g
AAJ6093606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6093603 Supplier Thermo Scientific Chemicals Supplier No. J6093603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 60-92-4 | C10H12N5O6P | 329.21 g/mol

cAMP is used for intracellular signal transduction in many different organisms, conveying the cAMP-dependent pathway. Restoration of several morphological characteristics of normal fibroblasts in sarcoma cells when treated with adenosine-3?: 5?-cyclic monophosphate. In the presence of adenosine 3?:5?-cyclic monophosphate, protein kinase I dissociated into two subunits: a subunit binding adenosine 3?:5?-cyclic monophosphate, and a catalytic subunit. Stimulation of calcium uptake in platelet membrane vesicles by adenosine 3?,5?-cyclic monophosphate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cAMP is used for intracellular signal transduction in many different organisms, conveying the cAMP-dependent pathway. Restoration of several morphological characteristics of normal fibroblasts in sarcoma cells when treated with adenosine-3′: 5′-cyclic monophosphate. In the presence of adenosine 3′:5′-cyclic monophosphate, protein kinase I dissociated into two subunits: a subunit binding adenosine 3′:5′-cyclic monophosphate, and a catalytic subunit. Stimulation of calcium uptake in platelet membrane vesicles by adenosine 3′,5′-cyclic monophosphate.

Solubility
Solube in water: 10 mg/mL pH of aqueous solution is approx. 3.0.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 60-92-4
Molecular Formula C10H12N5O6P
Molecular Weight (g/mol) 329.21
MDL Number MFCD00005845
InChI Key IVOMOUWHDPKRLL-YPLCUDRINA-N
Synonym camp, cyclic amp, adenosine 3',5'-cyclic monophosphate, 3',5'-cyclic amp, adenosine 3',5'-phosphate, adenosine 3',5'-cyclophosphate, cyclic 3',5'-amp, adenosine cyclic monophosphate, adenosine-3',5'-cyclophosphate, cyclic adenosine monophosphate
PubChem CID 6076
ChEBI CHEBI:17489
IUPAC Name (4aR,6R,7R,7aS)-6-(6-aminopurin-9-yl)-2-hydroxy-2-oxo-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-7-ol
SMILES NC1=C2N=CN([C@@H]3O[C@@H]4COP(O)(=O)O[C@H]4[C@H]3O)C2=NC=N1

Specifications

Melting Point 260°C (decomposition)
Color White
Quantity 1 g
Beilstein 52645
Merck Index 14,2708
Solubility Information Solube in water: 10mg/mL pH of aqueous solution is approx. 3.0.
Formula Weight 329.21
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Adenosine-3',5'-cyclic monophosphate
Hazard Category H314-H500
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 200-492-9
RTECSNumber AU7357600
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only