Learn More
Adamantane, 98%
CAS: 281-23-2 | C10H16 | 136.238 g/mol
Supplier: Thermo Scientific Chemicals A1404130
Description
Adamantane is mainly used for the synthesis of medicine to treat influenza, serve as NMDA receptor channel blockers. Also be used for preparation of advanced lubricants, photographic material, surface active agents, catalysts and other products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAdamantane is mainly used for the synthesis of medicine to treat influenza, serve as NMDA receptor channel blockers. Also be used for preparation of advanced lubricants, photographic material, surface active agents, catalysts and other products.
Solubility
Insoluble in water.
Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent.
Specifications
| Adamantane | |
| 266°C to 268°C (sealed tube) | |
| C10H16 | |
| 250 g | |
| 14,149 | |
| Insoluble in water. | |
| C1C2CC3CC1CC(C2)C3 | |
| 136.238 | |
| CHEBI:40519 | |
| 98% |
| 281-23-2 | |
| 1.07 | |
| MFCD00074719 | |
| 1901173 | |
| tricyclo 3.3.1.13,7 decane, adamantan, tricyclo 3.3.1.1 3,7 decane, tricyclo 3.3.1.1 decane, unii-pjy633525u, tricyclo 3.3.1.1∼3,7∼ decane, adamantine, 2-adamantyl, adamantan-1-yl | |
| ORILYTVJVMAKLC-UHFFFAOYSA-N | |
| adamantane | |
| 9238 | |
| 136.24 |
Chemical Identifiers
| 281-23-2 | |
| 136.238 | |
| ORILYTVJVMAKLC-UHFFFAOYSA-N | |
| 9238 | |
| adamantane |
| C10H16 | |
| MFCD00074719 | |
| tricyclo 3.3.1.13,7 decane, adamantan, tricyclo 3.3.1.1 3,7 decane, tricyclo 3.3.1.1 decane, unii-pjy633525u, tricyclo 3.3.1.1∼3,7∼ decane, adamantine, 2-adamantyl, adamantan-1-yl | |
| CHEBI:40519 | |
| C1C2CC3CC1CC(C2)C3 |
Safety and Handling
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 206-001-4
RTECSNumber : YK0493000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only