missing translation for 'onlineSavingsMsg'
Learn More

Adamantane, 98%

Catalog No. p-7021630
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1404118 50 g
AAA1404130 250 g
AAA140410B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1404118 Supplier Thermo Scientific Chemicals Supplier No. A1404118
Only null left

CAS: 281-23-2 | C10H16 | 136.238 g/mol

Adamantane is mainly used for the synthesis of medicine to treat influenza, serve as NMDA receptor channel blockers. Also be used for preparation of advanced lubricants, photographic material, surface active agents, catalysts and other products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adamantane is mainly used for the synthesis of medicine to treat influenza, serve as NMDA receptor channel blockers. Also be used for preparation of advanced lubricants, photographic material, surface active agents, catalysts and other products.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent.

Chemical Identifiers

CAS 281-23-2
Molecular Formula C10H16
Molecular Weight (g/mol) 136.238
MDL Number MFCD00074719
InChI Key ORILYTVJVMAKLC-UHFFFAOYSA-N
Synonym tricyclo 3.3.1.13,7 decane, adamantan, tricyclo 3.3.1.1 3,7 decane, tricyclo 3.3.1.1 decane, unii-pjy633525u, tricyclo 3.3.1.1∼3,7∼ decane, adamantine, 2-adamantyl, adamantan-1-yl
PubChem CID 9238
ChEBI CHEBI:40519
IUPAC Name adamantane
SMILES C1C2CC3CC1CC(C2)C3

Specifications

Melting Point 266°C to 268°C (sealed tube)
Density 1.07
Quantity 50 g
Beilstein 1901173
Merck Index 14,149
Solubility Information Insoluble in water.
Formula Weight 136.24
Percent Purity 98%
Chemical Name or Material Adamantane
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 206-001-4
RTECSNumber YK0493000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only