accessibility menu, dialog, popup

UserName

CAS RN 281-23-2

IUPAC Name: adamantane
Synonyms: adamantane tricyclo[3.3.1.1]decane
Molecular Weight (g/mol): 136.24
Molecular Formula: C10H16
InChi Key: ORILYTVJVMAKLC-UHFFFAOYSA-N
SMILES: C1C2CC3CC1CC(C2)C3
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)