Learn More
Fisherbrand™ ADA Height Wood Cabinet
Fisherbrand™ ADA Height Wood Cabinets bring quality, warmth and durability to your laboratory.
$3561.07 - $3772.66
Specifications
| Cabinet Type | ADA Height |
|---|---|
| Certifications/Compliance | SEFA 8 |
| Depth (English) | 23.375 in. |
| Depth (Metric) | 59.4 cm |
| Height (English) | 32.5 in. |
| Catalog Number | Mfr. No. | Color | Material | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Material | Price | Quantity | ||||
|
FB3644M1
|
N/A | Natural Stain | Maple Veneer |
Each for $3,772.66
|
|
||||
|
FB3644M2
|
N/A | Honey Stain | Maple Veneer |
Each for $3,772.66
|
|
||||
|
FB3644M3
|
N/A | Amber Stain | Maple Veneer |
Each for $3,772.66
|
|
||||
|
FB3644M4
|
N/A | Auburn Stain | Maple Veneer |
Each for $3,772.66
|
|
||||
|
FB3644M5
|
N/A | Harvest Stain | Maple Veneer |
Each for $3,772.66
|
|
||||
|
FB3644M7
|
N/A | Mocha Stain | Maple Veneer |
Each for $3,772.66
|
|
||||
|
FB3644O1
|
N/A | Natural Stain | Oak Veneer |
Each for $3,561.07
|
|
||||
|
FB3644O2
|
N/A | Honey Stain | Oak Veneer |
Each for $3,561.07
|
|
||||
|
FB3644O3
|
N/A | Amber Stain | Oak Veneer |
Each for $3,561.07
|
|
||||
|
FB3644O4
|
N/A | Auburn Stain | Oak Veneer |
Each for $3,561.07
|
|
||||
|
FB3644O5
|
N/A | Harvest Stain | Oak Veneer |
Each for $3,561.07
|
|
||||
|
FB3644O7
|
N/A | Mocha Stain | Oak Veneer |
Each for $3,561.07
|
|
||||
Description
- ADA Height (32.5 in. tall)
- Made from hand selected premium rotary cut maple or rift cut oak or rift cut oak veneers.
- Featuring solid mortise/tenon top frame, doweled, glued and screwed construction.
- Robotically applied environmentally friendly finishes are chemically resistant.
- Cabinets have a full flush overlay design with squared edges.
- They are vertically grain matched and are stained inside and out.
Compliance
SEFA 8
Specifications
| ADA Height | |
| 23.375 in. | |
| 32.5 in. | |
| 3 | |
| 18 in. | |
| ADA Height Cabinet |
| SEFA 8 | |
| 59.4 cm | |
| 82.6 cm | |
| 2 years | |
| 45.7 cm |