missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ ADA Height Wood Cabinet

Fisherbrand™ ADA Height Wood Cabinets bring quality, warmth and durability to your laboratory.

 View more versions of this product

Catalog No. FB3644O3


Only null left
Explore available promotions

Description

Description

  • ADA Height (32.5 in. tall)
  • Made from hand selected premium rotary cut maple or rift cut oak or rift cut oak veneers.
  • Featuring solid mortise/tenon top frame, doweled, glued and screwed construction.
  • Robotically applied environmentally friendly finishes are chemically resistant.
  • Cabinets have a full flush overlay design with squared edges.
  • They are vertically grain matched and are stained inside and out.

Compliance

SEFA 8

Specifications

Specifications

2 years
ADA Height Cabinet
82.6 cm
18 in.
45.7 cm
SEFA 8
Oak Veneer
3
23.375 in.
32.5 in.
59.4 cm
ADA Height
Amber Stain
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions