missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals ADA, 99%, for biochemistry
CAS: 26239-55-4 | C6H10N2O5 | 190.155 g/mol
$100.57 - $100.57
Chemical Identifiers
| CAS | 26239-55-4 |
|---|---|
| Molecular Formula | C6H10N2O5 |
| Molecular Weight (g/mol) | 190.155 |
| MDL Number | MFCD00008031 |
| InChI Key | QZTKDVCDBIDYMD-UHFFFAOYSA-N |
| Synonym | N-(2-Acetamido)iminodiacetic acid, N-(Carbamoylmethyl)iminodiacetic acid |
| PubChem CID | 117765 |
| ChEBI | CHEBI:43960 |
| IUPAC Name | 2-[(2-amino-2-oxoethyl)-(carboxymethyl)amino]acetic acid |
| SMILES | C(C(=O)N)N(CC(=O)O)CC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC228010250
|
Thermo Scientific Chemicals
228010250 |
25 g | Glass bottle |
Each for $100.57
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
| 26239-55-4 | |
| 190.155 | |
| QZTKDVCDBIDYMD-UHFFFAOYSA-N | |
| 117765 | |
| 2-[(2-amino-2-oxoethyl)-(carboxymethyl)amino]acetic acid |
| C6H10N2O5 | |
| MFCD00008031 | |
| N-(2-Acetamido)iminodiacetic acid, N-(Carbamoylmethyl)iminodiacetic acid | |
| CHEBI:43960 | |
| C(C(=O)N)N(CC(=O)O)CC(=O)O |
Specifications
| (9% soln. in 1 M NaOH), (9% soln. in 1 M NaOH) at 0nm, 0.08 max. at 300nm | |
| 219.0°C | |
| 98.5 | |
| White | |
| 25 g | |
| Authentic | |
| C6H10N2O5 | |
| MFCD00008031 | |
| 15, 135 | |
| Solubility in water: soluble. Other solubilities: soluble in alcohol, insoluble in benzene, chloroform | |
| C(C(=O)N)N(CC(=O)O)CC(=O)O | |
| 190.155 | |
| CHEBI:43960 | |
| 99% | |
| ADA, For biochemistry |
| (9% soln. in 1M NaOH) | |
| 26239-55-4 | |
| 100.0 | |
| Fine Crystalline Powder | |
| 99% | |
| 0.5% max. (110°C, 5 hrs) | |
| H2NCOCH2N(CH2CO2H)2 | |
| Glass bottle | |
| N-(2-Acetamido)iminodiacetic acid, N-(Carbamoylmethyl)iminodiacetic acid | |
| QZTKDVCDBIDYMD-UHFFFAOYSA-N | |
| 2-[(2-amino-2-oxoethyl)-(carboxymethyl)amino]acetic acid | |
| 117765 | |
| 190.16 | |
| Biochemical |
RUO – Research Use Only