missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals ADA, 99%, for biochemistry

Catalog No. AC228011000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC228011000 100 g Glass bottle
AC228010250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC228011000 Supplier Thermo Scientific Chemicals Supplier No. 228011000
Only null left

CAS: 26239-55-4 | C6H10N2O5 | 190.155 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer

Chemical Identifiers

CAS 26239-55-4
Molecular Formula C6H10N2O5
Molecular Weight (g/mol) 190.155
MDL Number MFCD00008031
InChI Key QZTKDVCDBIDYMD-UHFFFAOYSA-N
Synonym N-(2-Acetamido)iminodiacetic acid, N-(Carbamoylmethyl)iminodiacetic acid
PubChem CID 117765
ChEBI CHEBI:43960
IUPAC Name 2-[(2-amino-2-oxoethyl)-(carboxymethyl)amino]acetic acid
SMILES C(C(=O)N)N(CC(=O)O)CC(=O)O

Specifications

Absorbance (9% soln. in 1 M NaOH), (9% soln. in 1 M NaOH) at 0nm, 0.08 max. at 300nm
Absorbance Conditions (9% soln. in 1M NaOH)
Melting Point 219.0°C
CAS Min % 98.5
Color White
CAS Max % 100.0
Physical Form Fine Crystalline Powder
Quantity 100 g
Assay Percent Range 99%
Infrared Spectrum Authentic
Loss on Drying 0.5% max. (110°C, 5 hrs)
Linear Formula H2NCOCH2N(CH2CO2H)2
Packaging Glass bottle
Merck Index 15, 135
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, insoluble in benzene, chloroform
Formula Weight 190.16
Percent Purity 99%
Grade Biochemical
Chemical Name or Material ADA, For biochemistry
Show More Show Less

RUO – Research Use Only