missing translation for 'onlineSavingsMsg'
Learn More

Actinomycin D (1 mg/ml, DMSO)

CAS: 50-76-0 | C62H86N12O16 | 1255.438 g/mol

Supplier:  Thermo Scientific Chemicals J67160XF

 View more versions of this product

Catalog No. AAJ67160XF

Only null left

Description

Description

Actinomycin D has a wide-ranging applications ranging from a selection agent in cell culture, use in studies of suppressing HIV-replication and programmed cell death of PC12 cells, and as an antibiotic in treatment of various malignant neoplasms, including Wilm?s tumor and the sarcomas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Actinomycin D has a wide-ranging applications ranging from a selection agent in cell culture, use in studies of suppressing HIV-replication and programmed cell death of PC12 cells, and as an antibiotic in treatment of various malignant neoplasms, including Wilm′s tumor and the sarcomas.

Solubility
Soluble in acetonitrilie or acetone at 10 mg/mL and in DMSO at a minimum of 1 mg/mL, slightly soluble in water.

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Specifications

Specifications

Actinomycin D
1mg/mL in DMSO, sterile-filtered
50-76-0
C62H86N12O16
Dactinomycin; Actinomycin C1
RJURFGZVJUQBHK-HUZQGMAJSA-N
2-amino-4,6-dimethyl-3-oxo-1-N-[(3R,6S,7R,10S,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]-9-N-[(3R,6S,7R,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,
131954673
Actinomycin D
Liquid
1 mL
MFCD00005033
Soluble in acetonitrilie or acetone at 10mg/mL and in DMSO at a minimum of 1mg/mL,slightly soluble in water.
CC1C(C(=O)NC(C(=O)N2CCCC2C(=O)N(CC(=O)N(C(C(=O)O1)C(C)C)C)C)C(C)C)NC(=O)C3=C4C(=C(C=C3)C)OC5=C(C(=O)C(=C(C5=N4)C(=O)NC6C(OC(=O)C(N(C(=O)CN(C(=O)C7CCCN7C(=O)C(NC6=O)C(C)C)C)C)C(C)C)C)N)C
1255.438

Chemical Identifiers

50-76-0
1255.438
RJURFGZVJUQBHK-HUZQGMAJSA-N
131954673
CC1C(C(=O)NC(C(=O)N2CCCC2C(=O)N(CC(=O)N(C(C(=O)O1)C(C)C)C)C)C(C)C)NC(=O)C3=C4C(=C(C=C3)C)OC5=C(C(=O)C(=C(C5=N4)C(=O)NC6C(OC(=O)C(N(C(=O)CN(C(=O)C7CCCN7C(=O)C(NC6=O)C(C)C)C)C)C(C)C)C)N)C
C62H86N12O16
MFCD00005033
Dactinomycin; Actinomycin C1
2-amino-4,6-dimethyl-3-oxo-1-N-[(3R,6S,7R,10S,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]-9-N-[(3R,6S,7R,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 200-063-6

RTECSNumber : AU1575000

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only