missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acrylamide, ≥99%, MP Biomedicals™
Acrylamide, ≥99%, White Crystalline Powder
$124.06 - $187.79
Chemical Identifiers
| CAS | 79-06-1 |
|---|---|
| Molecular Formula | C3H5NO |
| Molecular Weight (g/mol) | 71.08 |
| MDL Number | MFCD00008032 |
| InChI Key | HRPVXLWXLXDGHG-UHFFFAOYSA-N |
| Synonym | 2-Propenamide, Acrylic acid amide |
| PubChem CID | 6579 |
| ChEBI | CHEBI:28619 |
| IUPAC Name | prop-2-enamide |
| SMILES | NC(=O)C=C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15025680
|
MP Biomedicals Inc
0215025680 |
100 g |
Each for $124.06
|
|
|||||
|
ICN15025690
|
MP Biomedicals Inc
0215025690 |
500 g |
Each for $187.79
|
|
|||||
Description
- Acrylamide is a monomer used in a variety of synthetic processes to form polymers and copolymers.
- It polymerizes in the presence of free radicals in aqueous solutions.
- It is usually used to prepare polyacrylamide gels for electrophoresis separation of biomolecules.
- Acrylamide is prepared on an industrial scale by the hydrolysis of acrylonitrile by nitrile hydratase.
Chemical Identifiers
| 79-06-1 | |
| 71.08 | |
| HRPVXLWXLXDGHG-UHFFFAOYSA-N | |
| 6579 | |
| prop-2-enamide |
| C3H5NO | |
| MFCD00008032 | |
| 2-Propenamide, Acrylic acid amide | |
| CHEBI:28619 | |
| NC(=O)C=C |
Specifications
| Crystalline Powder | |
| 1.127g/cm3 at 25°C | |
| 100 g | |
| C3H5NO | |
| 2-Propenamide, Acrylic acid amide | |
| NC(=O)C=C | |
| 71.08 | |
| CHEBI:28619 | |
| Acrylamide |
| 79-06-1 | |
| White | |
| ≥99% | |
| MFCD00008032 | |
| HRPVXLWXLXDGHG-UHFFFAOYSA-N | |
| prop-2-enamide | |
| 6579 | |
| 71.1 |