missing translation for 'onlineSavingsMsg'
Learn More

Acrylamide, ≥99%, MP Biomedicals™

Acrylamide, ≥99%, White Crystalline Powder

Supplier:  MP Biomedicals Inc 0215025690

 View more versions of this product

Catalog No. ICN15025690


Only null left
Explore available promotions

Description

Description

  • Acrylamide is a monomer used in a variety of synthetic processes to form polymers and copolymers.
  • It polymerizes in the presence of free radicals in aqueous solutions.
  • It is usually used to prepare polyacrylamide gels for electrophoresis separation of biomolecules.
  • Acrylamide is prepared on an industrial scale by the hydrolysis of acrylonitrile by nitrile hydratase.
Specifications

Specifications

Acrylamide
79-06-1
White
≥99%
MFCD00008032
HRPVXLWXLXDGHG-UHFFFAOYSA-N
prop-2-enamide
6579
71.1
Crystalline Powder
1.127g/cm3 at 25°C
500 g
C3H5NO
2-Propenamide, Acrylic acid amide
NC(=O)C=C
71.08
CHEBI:28619

Chemical Identifiers

79-06-1
71.08
HRPVXLWXLXDGHG-UHFFFAOYSA-N
6579
prop-2-enamide
C3H5NO
MFCD00008032
2-Propenamide, Acrylic acid amide
CHEBI:28619
NC(=O)C=C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions