missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acrylamide, ≥99%, MP Biomedicals™
Acrylamide, ≥99%, White Crystalline Powder
Supplier: MP Biomedicals Inc 0215025680
Description
- Acrylamide is a monomer used in a variety of synthetic processes to form polymers and copolymers.
- It polymerizes in the presence of free radicals in aqueous solutions.
- It is usually used to prepare polyacrylamide gels for electrophoresis separation of biomolecules.
- Acrylamide is prepared on an industrial scale by the hydrolysis of acrylonitrile by nitrile hydratase.
Specifications
| Acrylamide | |
| 79-06-1 | |
| White | |
| ≥99% | |
| MFCD00008032 | |
| HRPVXLWXLXDGHG-UHFFFAOYSA-N | |
| prop-2-enamide | |
| 6579 | |
| 71.1 |
| Crystalline Powder | |
| 1.127g/cm3 at 25°C | |
| 100 g | |
| C3H5NO | |
| 2-Propenamide, Acrylic acid amide | |
| NC(=O)C=C | |
| 71.08 | |
| CHEBI:28619 |
Chemical Identifiers
| 79-06-1 | |
| 71.08 | |
| HRPVXLWXLXDGHG-UHFFFAOYSA-N | |
| 6579 | |
| prop-2-enamide |
| C3H5NO | |
| MFCD00008032 | |
| 2-Propenamide, Acrylic acid amide | |
| CHEBI:28619 | |
| NC(=O)C=C |