missing translation for 'onlineSavingsMsg'
Learn More

Acrolein Standard, 5000 μg/mL in P&T methanol, Restek

Catalog No. 06722501
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06722501 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06722501 Supplier Chromatographic Supplier No. RK 30645

Chemical Identifiers

CAS 107-02-8 , 67-56-1
Molecular Formula C3H4O
Molecular Weight (g/mol) 56.06
MDL Number MFCD00006998
InChI Key HGINCPLSRVDWNT-UHFFFAOYSA-N
PubChem CID 7847
ChEBI CHEBI:15368
IUPAC Name prop-2-enal
SMILES C=CC=O

Specifications

Density 1.1 (air = 1) vapour
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Methanol, acrolein
Chemical Name or Material Acrolein Standard
ShelfLife Limited shelf life
Recommended Storage Store in a cool dry ventilated location. Keep container(s) closed away from sources of ignition.