CAS RN 107-02-8
IUPAC Name:
prop-2-enal
Synonyms:
acrolein caswell no. 009 crolean acrolein (substance) magnacide H akrolein acquinite rcra waste number P003 acroleina acroleine
Molecular Weight (g/mol):
56.06
Molecular Formula:
C3H4O
InChi Key:
HGINCPLSRVDWNT-UHFFFAOYSA-N
SMILES:
C=CC=O
1
–
1
of
1
results
1
Acrolein Standard, 5000 μg/mL in P&T methanol, Restek
CAS: 107-02-8 Molecular Formula: C3H4O Molecular Weight (g/mol): 56.06 MDL Number: MFCD00006998 InChI Key: HGINCPLSRVDWNT-UHFFFAOYSA-N PubChem CID: 7847 ChEBI: CHEBI:15368 IUPAC Name: prop-2-enal SMILES: C=CC=O