missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acid Alcohol, Strong, 5% (v/v) Hydrochloric Acid in 95% Denatured Alcohol, Ricca Chemical
$82.78 - $82.78
Chemical Identifiers
| CAS | 64-17-5 |
|---|---|
| InChI Key | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
| PubChem CID | 702 |
| ChEBI | CHEBI:16236 |
Chemical Identifiers
| 64-17-5 | |
| LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
| 702 | |
| CHEBI:16236 |
Specifications
| Solution | |
| 94.0,4.0 | |
| Natural Poly Bottle | |
| <1 | |
| LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
| CHEBI:16236 | |
| 5% (v/v) Hydrochloric Acid in 95% Denatured Alcohol |
| 64-17-5,7732-18-5,67-56-1,7647-01-0 | |
| 96.0,6.0 | |
| Colorless | |
| 500 mL | |
| 702 | |
| Laboratory | |
| Acid Alcohol, Strong |