missing translation for 'onlineSavingsMsg'
Learn More

Acid Alcohol, Strong, 5% (v/v) Hydrochloric Acid in 95% Denatured Alcohol, Ricca Chemical

Supplier:  Ricca Chemical Company RI26016

 View more versions of this product

Catalog No. 260-16


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Solution
64-17-5,7732-18-5,67-56-1,7647-01-0
96.0,6.0
Colorless
500 mL
702
Laboratory
Acid Alcohol, Strong
94.0,4.0
Natural Poly Bottle
<1
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
CHEBI:16236
5% (v/v) Hydrochloric Acid in 95% Denatured Alcohol

Chemical Identifiers

64-17-5
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
702
CHEBI:16236
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions