missing translation for 'onlineSavingsMsg'
Learn More

Acetyl chloride, 1M solution in dichloromethane, AcroSeal™

CAS: 75-36-5 | C2H3ClO | 78.50 g/mol

$130.18 - $130.18

Specifications

Chemical Name or Material Acetyl chloride
Color Colorless
Name Note 1M solution in dichloromethane
CAS 75-36-5 , 75-09-2
CAS Min % 5.44,93.36
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC387741000
View Documents
Thermo Scientific Chemicals
387741000
100 mL AcroSeal™ Glass bottle
Each for $130.18
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Acetyl chloride
1M solution in dichloromethane
5.44,93.36
C2H3ClO
MFCD00000719
1.3000g/mL
ethanoyl chloride, acetic chloride, acetylchloride, acetic acid chloride, ch3cocl, rcra waste number u006, acetic acid, chloride, unii-qd15rno45k, ccris 4568, hsdb 662
WETWJCDKMRHUPV-UHFFFAOYSA-N
acetyl chloride
6367
78.5
Liquid
Colorless
75-36-5 , 75-09-2
6.64,94.57
CH3COCl
0.9 to 1.1M
15, 80
Solubility in water: reacts with water.
CC(Cl)=O
78.50
CHEBI:37580
1.3
Videos
SDS
Documents

Documents

RUO – Research Use Only