missing translation for 'onlineSavingsMsg'
Learn More

Acetrizoic Acid 98.0+%, TCI America™

Catalog No. A08585G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A08585G 5 g
A08581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A08585G Supplier TCI America Supplier No. A08585G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 85-36-9
Molecular Formula C9H6I3NO3
Molecular Weight (g/mol) 556.864
MDL Number MFCD00016495
InChI Key GNOGSFBXBWBTIG-UHFFFAOYSA-N
Synonym acetrizoic acid, urokon, opacaron, acetrizoesaeure, urokonic acid, acide acetrizoique, acetriazoic acid, 3-acetylamino-2,4,6-triiodobenzoic acid, unii-24256bqv7m, benzoic acid, 3-acetylamino-2,4,6-triiodo
PubChem CID 6806
ChEBI CHEBI:34521
IUPAC Name 3-acetamido-2,4,6-triiodobenzoic acid
SMILES CC(=O)NC1=C(C=C(C(=C1I)C(=O)O)I)I

Specifications

Color White
Quantity 5 g
Formula Weight 556.86
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Acetrizoic Acid
RTECSNumber DG0887000
TSCA No