accessibility menu, dialog, popup

UserName

CH3ONHIIHOOI

CAS RN 85-36-9

IUPAC Name: 3-acetamido-2,4,6-triiodobenzoic acid
Synonyms: acetrizoic acid
Molecular Weight (g/mol): 556.86
Molecular Formula: C9H6I3NO3
InChi Key: GNOGSFBXBWBTIG-UHFFFAOYSA-N
SMILES: CC(=O)NC1=C(I)C=C(I)C(C(O)=O)=C1I
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results