missing translation for 'onlineSavingsMsg'
Learn More

Acetonyltriphenylphosphonium bromide, 98+%

Catalog No. AAB2243406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2243406 5 g
AAB2243414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2243406 Supplier Thermo Scientific Chemicals Supplier No. B2243406

CAS: 2236-01-3 | C21H20BrOP | 399.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2236-01-3
Molecular Formula C21H20BrOP
Molecular Weight (g/mol) 399.27
MDL Number MFCD00051863
InChI Key KZBWHCFTAAHIJJ-UHFFFAOYSA-M
Synonym acetonyltriphenylphosphonium bromide, acetonyl triphenylphosphonium bromide, 2-oxopropyl triphenylphosphanium bromide, phosphonium, 2-oxopropyl triphenyl-, bromide, acetonyltriphenylphosphoniumbromide, acmc-1cmpe, 2-oxopropyl-triphenylphosphonium bromide, 2-oxopropyl triphenylphosphonium bromide, 2-oxopropyl triphenyl phosphanium bromide, phosphonium, 2-oxopropyl triphenyl-,bromide 1:1
PubChem CID 638158
IUPAC Name 2-oxopropyl(triphenyl)phosphanium;bromide
SMILES [Br-].CC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 226°C to 227°C
Quantity 5 g
Beilstein 3802148
Sensitivity Hygroscopic
Formula Weight 399.27
Percent Purity ≥98%
Chemical Name or Material Acetonyltriphenylphosphonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-796-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only