missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, R CHROMASOLV™, for liquid chromatography, ≥99.8% (GC), Honeywell Riedel-de Haën™
R CHROMASOLV™, for liquid chromatography,≥99.8% (GC)
$305.55 - $3186.70
Chemical Identifiers
| CAS | 75-05-8 |
|---|---|
| Molecular Formula | C2H3N |
| Molecular Weight (g/mol) | 41.053 |
| MDL Number | MFCD00001878 |
| InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
| Synonym | methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn |
| PubChem CID | 6342 |
| ChEBI | CHEBI:38472 |
| IUPAC Name | acetonitrile |
| SMILES | CC#N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
6000255
|
Honeywell-Chromasolv
348811L |
1 L | Glass Bottle |
Each for $305.55
|
|
||||
|
6000256
|
Honeywell Chemicals
3488125L |
2.5 L | Glass Bottle |
Each for $589.70
|
|
||||
|
6000257
|
Honeywell-Chromasolv
348816X1L |
6 x 1 L | Glass Bottle |
Case of 6 Each for $1,563.90
|
|
||||
|
60046500
|
Honeywell Chemicals
348814X25L |
4 x 2.5 L | Glass Bottle |
Case of 4 Each for $1,967.33
|
|
||||
|
6000260
|
Honeywell Chemicals
348814X4L |
4 x 4 L | Glass Bottle |
Case of 4 Each for $3,186.70
|
|
||||
Description
- For GC headspace applications
- Meets USP, Ph. Eur. and ICH requirements
- GC headspace application tested
- Packed under inert gas for longer shelf life
Chemical Identifiers
| 75-05-8 | |
| 41.053 | |
| WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
| 6342 | |
| acetonitrile |
| C2H3N | |
| MFCD00001878 | |
| methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
| CHEBI:38472 | |
| CC#N |
Specifications
| -48°C (literature) | |
| 100.0000% | |
| 81.6°C | |
| 6°C | |
| C2H3N | |
| MFCD00001878 | |
| 741857 | |
| WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
| acetonitrile | |
| 6342 | |
| 41.05 | |
| Chromatography |
| 75-05-8 | |
| 0.786Â g/mLÂ at 25Â °C(lit.) | |
| 1 L | |
| Glass Bottle | |
| CH3CN | |
| UN1648 | |
| methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
| CC#N | |
| 41.053 | |
| CHEBI:38472 | |
| >99.8% | |
| Acetonitrile, R CHROMASOLV™, for liquid chromatography, ≥99.8% (GC) |
Safety and Handling
ShelfLife : 1260 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile
EINECSNumber : 200-835-2
Recommended Storage : Room Temp