missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, R CHROMASOLV™, for liquid chromatography, ≥99.8% (GC), Solstice

Catalog No. 6000260
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000260 4 x 4 L Glass Bottle
6000255 1 L Glass Bottle
6000256 2.5 L Glass Bottle
6000257 6 x 1 L Glass Bottle
60046500 4 x 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 6000260 Supplier Solstice Supplier No. 348814X4L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R CHROMASOLV™, for liquid chromatography,≥99.8% (GC)

  • For GC headspace applications
  • Meets USP, Ph. Eur. and ICH requirements
  • GC headspace application tested
  • Packed under inert gas for longer shelf life

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -48°C (literature)
CAS Max % 100.0000%
Density 0.786 g/mL at 25 °C(lit.)
Quantity 4 x 4 L
Flash Point 6°C
Packaging Glass Bottle
Linear Formula CH3CN
UN Number UN1648
Beilstein 741857
Boiling Point 81.6°C
Formula Weight 41.05
Percent Purity >99.8%
Grade Chromatography
Chemical Name or Material Acetonitrile, R CHROMASOLV™, for liquid chromatography, ≥99.8% (GC)
Show More Show Less
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile
EINECSNumber 200-835-2
Recommended Storage Room Temp