missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, R CHROMASOLV™, for liquid chromatography, ≥99.8% (GC), Honeywell Riedel-de Haën™
R CHROMASOLV™, for liquid chromatography,≥99.8% (GC)
Supplier: Honeywell-Chromasolv 3488125L
Description
- For GC headspace applications
- Meets USP, Ph. Eur. and ICH requirements
- GC headspace application tested
- Packed under inert gas for longer shelf life
Specifications
| Acetonitrile | |
| R CHROMASOLV™, for liquid chromatography, ≥99.8% (GC) | |
| 100.0000% | |
| 81.6°C | |
| 6°C | |
| C2H3N | |
| MFCD00001878 | |
| 741857 | |
| WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
| acetonitrile | |
| 6342 | |
| 41.05 | |
| Chromatography |
| -48°C (literature) | |
| 75-05-8 | |
| 0.786Â g/mLÂ at 25Â °C(lit.) | |
| 2.5 L | |
| Glass Bottle | |
| CH3CN | |
| UN1648 | |
| methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
| CC#N | |
| 41.053 | |
| CHEBI:38472 | |
| >99.8% |
Chemical Identifiers
| 75-05-8 | |
| 41.053 | |
| WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
| 6342 | |
| acetonitrile |
| C2H3N | |
| MFCD00001878 | |
| methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
| CHEBI:38472 | |
| CC#N |
Safety and Handling
ShelfLife : 1260 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile
EINECSNumber : 200-835-2
Recommended Storage : Room Temp