missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone, Puriss., meets analytical spec. of BP, NF, Ph. Eur., ≥99% (GC), Honeywell Riedel-de Haën™
Puriss., meets analytical specification of BP, NF, Ph. Eur.,≥99% (GC)
Supplier: Honeywell Inc 242014X25LGL
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
| Acetone | |
| Meets analytical specification of Ph. Eur.BP, NF | |
| Colorless | |
| 55°C to 57°C | |
| 0.132 | |
| Glass Bottle | |
| C3H6O | |
| MFCD00008765 | |
| UN1090 | |
| acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
| CC(C)=O | |
| 58.08 | |
| CHEBI:15347 | |
| ≥99% (GC) |
| -95°C | |
| 67-64-1 | |
| 0.791g/mL at 25°C, 0.790 to 0.792g/mL at 20°C | |
| 4 x 2.5 L | |
| −17°C (1.4°F) | |
| Liquid | |
| CH3COCH3 | |
| 184mmHg (20°C) | |
| 635680 | |
| CSCPPACGZOOCGX-UHFFFAOYSA-N | |
| propan-2-one | |
| 180 | |
| 58.08g/mol | |
| puriss. |
Chemical Identifiers
| 67-64-1 | |
| 58.08 | |
| CSCPPACGZOOCGX-UHFFFAOYSA-N | |
| 180 | |
| propan-2-one |
| C3H6O | |
| MFCD00008765 | |
| acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
| CHEBI:15347 | |
| CC(C)=O |
Safety and Handling
P210-P305 + P351 + P338-P370 + P378-P403 + P235
H225-H319-H336
DOTInformation : Class 3 - PG 2 - Acetone
EINECSNumber : 200-662-2
RTECSNumber : AL3150000