missing translation for 'onlineSavingsMsg'
Learn More

Acetone, Puriss., meets analytical spec. of BP, NF, Ph. Eur., ≥99% (GC), Honeywell Riedel-de Haën™

Catalog No. p-7120105
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
155 kg
2.5 L
20 kg
4 x 2.5 L
4 x 5 L
5 L
6 x 1 L
Packaging:
Glass Bottle
Metal Drum
Poly Bottle
Stainless Steel Can
Catalog No. Quantity Packaging
60045553 1 L Poly Bottle
60045555 2.5 L Glass Bottle
60045554 2.5 L Poly Bottle
60045561 5 L Poly Bottle
60045562 6 x 1 L Poly Bottle
60045559 4 x 2.5 L Glass Bottle
60045558 4 x 2.5 L Poly Bottle
60045556 20 kg Stainless Steel Can
60045560 4 x 5 L Poly Bottle
60045552 155 kg Metal Drum
10 options

Catalog No. 60045553

Supplier: Honeywell Inc 242011L

Only null left

Puriss., meets analytical specification of BP, NF, Ph. Eur.,≥99% (GC)

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -95°C
Color Colorless
Density 0.791g/mL at 25°C, 0.790 to 0.792g/mL at 20°C
Boiling Point 55°C to 57°C
Quantity 1 L
Explosion Limit 0.132
Flash Point −17°C (1.4°F)
Packaging Poly Bottle
Physical Form Liquid
Linear Formula CH3COCH3
Vapor Pressure 184mmHg (20°C)
UN Number UN1090
Beilstein 635680
Formula Weight 58.08g/mol
Percent Purity ≥99% (GC)
Grade puriss.
Chemical Name or Material Acetone, Meets analytical specification of Ph. Eur.BP, NF
Show More Show Less
Hazard Statement P210-P305 + P351 + P338-P370 + P378-P403 + P235
Precautionary Statement H225-H319-H336
DOTInformation Class 3 - PG 2 - Acetone
EINECSNumber 200-662-2
RTECSNumber AL3150000