missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone, Purified Suitable for Histology, Reagents
Acetone, Purified Suitable for Histology, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C210250020E7
Specifications
| Acetone, Purified Suitable for Histology | |
| 67-64-1 | |
| 56°C | |
| HDPE | |
| 0.788 | |
| MFCD00008765 | |
| CSCPPACGZOOCGX-UHFFFAOYSA-N | |
| propan-2-one | |
| Pure |
| -94.7°C | |
| Colorless | |
| 20 L | |
| Liquid | |
| C3H6O | |
| Dimethyl Ketone, 2-Propanone | |
| CC(C)=O | |
| 58.08 |
Chemical Identifiers
| 67-64-1 | |
| 58.08 | |
| CSCPPACGZOOCGX-UHFFFAOYSA-N | |
| propan-2-one |
| C3H6O | |
| MFCD00008765 | |
| Dimethyl Ketone, 2-Propanone | |
| CC(C)=O |