missing translation for 'onlineSavingsMsg'
Learn More

Acetone, Purified Suitable for Histology, Reagents

Catalog No. p-200020878
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
200 L
Packaging:
Glass Bottle
HDPE
HDPE Winpak
Poly Bottle
Steel Drum
Steel Pail
Tin Can
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01337620 10 L Steel Pail
01337619 1 L Glass Bottle
01337618 200 L Steel Drum
01337617 20 L HDPE Winpak
01337616 20 L Steel Pail
01337615 20 L HDPE
01337614 4 L Poly Bottle
01337613 4 L Tin Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 01337620 Supplier Labchem Inc Supplier No. C210250010E6
Only null left

Acetone, Purified Suitable for Histology, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym Dimethyl Ketone, 2-Propanone
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -94.7°C
Color Colorless
Boiling Point 56°C
Quantity 10 L
Packaging Steel Pail
Physical Form Liquid
Specific Gravity 0.788
Grade Pure
Chemical Name or Material Acetone, Purified Suitable for Histology