missing translation for 'onlineSavingsMsg'
Learn More

Acetone (OmniSolv™) 99.5%, MilliporeSigma™

Supplier:  Technidata AX01161

 View more versions of this product

Catalog No. MAX01161


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Acetone, 99.5%
0.005AU max at 350nm, 0.005AU max at 400nm, 0.06AU max at 340nm, 0.30AU max at 335nm, 0.80AU max at 330nm, 1.00AU max at 329nm
4 L
Glass Bottle
0.3μeq/g max
CH3COCH3
0.5% max
CSCPPACGZOOCGX-UHFFFAOYSA-N
propan-2-one
180
58.08
OmniSolv
67-64-1
0.79kg/L
To Pass Test
1ppm max
C3H6O
MFCD00008765
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CC(C)=O
58.08
CHEBI:15347
≥99.5% (GC, corrected for water content)

Chemical Identifiers

67-64-1
58.08
CSCPPACGZOOCGX-UHFFFAOYSA-N
180
propan-2-one
C3H6O
MFCD00008765
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CHEBI:15347
CC(C)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions