missing translation for 'onlineSavingsMsg'
Learn More

Acetone (OmniSolv™) 99.5%, MilliporeSigma™

Catalog No. p-325411
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MAX01166 1 L Glass Bottle
MAX01161 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MAX01166 Supplier Technidata Supplier No. AX01166
Only null left

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Absorbance 0.005AU max at 350nm, 0.005AU max at 400nm, 0.06AU max at 340nm, 0.30AU max at 335nm, 0.80AU max at 330nm, 1.00AU max at 329nm
Density 0.79kg/L
Quantity 1 L
Infrared Spectrum To Pass Test
Packaging Glass Bottle
Residue after Evaporation 1ppm max
Titratable Acid 0.3μeq/g max
Linear Formula CH3COCH3
Water 0.5% max
Formula Weight 58.08
Percent Purity ≥99.5% (GC, corrected for water content)
Grade OmniSolv
Chemical Name or Material Acetone, 99.5%
Show More Show Less
Recommended Storage Room Temperature